Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 21 | 3534 | ||
NM_022489.3 | NP_071934.3 | 21 | 3534 | |||
XM_005268004.4 | XP_005268061.1 | 21 | 3621 | |||
XM_005268005.4 | XP_005268062.1 | 21 | 3592 | |||
XM_017021595.1 | XP_016877084.1 | 21 | 3592 | |||
GenBank mRNA | AF318379.1 | - | 3 | 617 | ||
AK025709.1 | - | 12 | 1497 | |||
AK225325.1 | - | 12 | 1497 | |||
BC008756.2 | - | 11 | 1442 | |||
BC064828.1 | - | 5 | 852 | |||
DQ395338.1 | - | 14 | 1795 | |||
DQ395339.1 | - | 14 | 1795 | |||
DQ395340.1 | - | 14 | 1795 |