Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080479.2 | NP_001073948.2 | 37 | 5606 | ||
NM_001177693.1 | NP_001171164.1 | 36 | 5528 | |||
NM_001244364.1 | NP_001231293.1 | 28 | 4484 | |||
XM_005248568.4 | XP_005248625.1 | 29 | 6043 | |||
XM_011543546.1 | XP_011541848.1 | 37 | 5613 | |||
XM_011543547.2 | XP_011541849.1 | 36 | 5840 | |||
XM_017009702.1 | XP_016865191.1 | 36 | 5291 | |||
XM_017009703.1 | XP_016865192.1 | 28 | 4344 | |||
GenBank mRNA | AK025470.1 | - | 12 | 2190 | ||
BC157846.1 | - | 36 | 5452 | 9057065 | ||
BC171850.1 | - | 35 | 5374 | 9054504 | ||
BG696752.1 | - | 1 | 99 |