Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022492.4 | NP_071937.4 | 13 | 1817 | ||
NR_027749.1 | 12 | 1708 | ||||
XM_005264492.1 | XP_005264549.1 | 13 | 1800 | |||
XM_011533035.1 | XP_011531337.1 | 13 | 1866 | |||
XM_011533036.1 | XP_011531338.1 | 13 | 1863 | |||
XM_011533037.1 | XP_011531339.1 | 13 | 1860 | |||
XM_011533040.1 | XP_011531342.1 | 12 | 1815 | |||
XM_017004706.1 | XP_016860195.1 | 13 | 1803 | |||
XM_017004707.1 | XP_016860196.1 | 13 | 1797 | |||
XM_017004708.1 | XP_016860197.1 | 12 | 1711 | |||
XM_017004709.1 | XP_016860198.1 | 12 | 1935 | |||
GenBank mRNA | AK022850.1 | - | 9 | 1327 | ||
AK026819.1 | - | 1 | 803 | |||
AK301585.1 | - | 6 | 872 | |||
AK303245.1 | - | 5 | 799 | |||
BC036017.1 | - | 13 | 1801 | 5270575 | ||
BC045536.1 | - | 12 | 1699 | |||
BX648131.1 | - | 7 | 2753 |