Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 15 | 2026 | ||
XM_006716381.2 | XP_006716444.1 | 15 | 2056 | |||
XM_006716385.3 | XP_006716448.1 | 16 | 3551 | |||
XM_017013732.1 | XP_016869221.1 | 15 | 2023 | |||
XM_017013733.1 | XP_016869222.1 | 16 | 2143 | |||
XM_017013734.1 | XP_016869223.1 | 15 | 2007 | |||
GenBank mRNA | AF495722.1 | - | 10 | 1433 | ||
AK025454.1 | - | 4 | 436 | |||
AK055191.1 | - | 9 | 1085 | |||
AK095924.1 | - | 2 | 639 | |||
AK294646.1 | - | 3 | 250 | |||
AK315634.1 | - | 3 | 292 | |||
BC012865.1 | - | 6 | 839 | |||
BC013350.2 | - | 5 | 517 | |||
BC052237.1 | - | 15 | 2023 | |||
BC112147.1 | - | 9 | 1115 |