Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MESDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022566.2 | NP_072088.1 | 1 | 2378 | ||
GenBank mRNA | AY007810.1 | - | 1 | 1700 | ||
BC084554.1 | - | 1 | 1237 | 6088754 |