Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022735.3 | NP_073572.2 | 3 | 520 | ||
GenBank mRNA | AB043587.2 | - | 3 | 528 | ||
AK025520.1 | - | 3 | 521 | |||
AK314468.1 | - | 3 | 521 | |||
AY150218.1 | - | 3 | 499 | |||
BC045533.1 | - | 3 | 506 | 5259930 | ||
BC060792.1 | - | 3 | 516 | 4813993 |