Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 17 | 2474 | ||
XM_006716381.2 | XP_006716444.1 | 17 | 2504 | |||
XM_006716385.3 | XP_006716448.1 | 18 | 3999 | |||
XM_017013732.1 | XP_016869221.1 | 17 | 2471 | |||
XM_017013733.1 | XP_016869222.1 | 18 | 2591 | |||
XM_017013734.1 | XP_016869223.1 | 17 | 2455 | |||
GenBank mRNA | AF318332.1 | - | 1 | 167 | ||
AF495722.1 | - | 12 | 1880 | |||
AK025411.1 | - | 3 | 448 | |||
AK025454.1 | - | 6 | 884 | |||
AK055191.1 | - | 11 | 1533 | |||
AK095924.1 | - | 4 | 1087 | |||
AK294646.1 | - | 5 | 698 | |||
BC012865.1 | - | 8 | 1287 | |||
BC013350.2 | - | 7 | 965 | |||
BC052237.1 | - | 17 | 2471 |