Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005435.3 | NP_005426.2 | 15 | 5367 | ||
GenBank mRNA | AK090570.1 | - | 14 | 2817 | ||
AK123596.1 | - | 14 | 3566 | |||
AK160365.1 | - | 15 | 5423 | |||
BC010046.2 | - | 15 | 2312 | |||
BC011839.2 | - | 15 | 2312 | |||
BC014555.2 | - | 15 | 2312 | |||
BX649178.1 | - | 14 | 3252 | |||
U02082.1 | - | 13 | 2107 |