Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022772.3 | NP_073609.2 | 4 | 362 | ||
XM_017018130.1 | XP_016873619.1 | 5 | 579 | |||
XM_017018131.1 | XP_016873620.1 | 4 | 450 | |||
XM_017018132.1 | XP_016873621.1 | 5 | 420 | |||
GenBank mRNA | AK025588.1 | - | 4 | 280 | ||
AK025824.1 | - | 4 | 280 | |||
AK122903.1 | - | 1 | 558 | |||
AK222903.1 | - | 4 | 275 | |||
AK225311.1 | - | 4 | 280 | |||
AK294041.1 | - | 4 | 279 | |||
AY074929.1 | - | 4 | 387 | |||
BC080636.1 | - | 4 | 366 | 6572993 | ||
BC093878.1 | - | 3 | 155 | 7939723 | ||
BC101481.1 | - | 3 | 155 | 8068987 | ||
BC143242.1 | - | 3 | 155 | 9051743 |