Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022662.3 | NP_073153.1 | 35 | 4605 | ||
XM_011511633.2 | XP_011509935.1 | 35 | 4649 | |||
XM_011511634.2 | XP_011509936.1 | 35 | 4764 | |||
XM_017004710.1 | XP_016860199.1 | 35 | 4649 | |||
XM_017004711.1 | XP_016860200.1 | 35 | 4637 | |||
XM_017004712.1 | XP_016860201.1 | 35 | 4752 | |||
XM_017004713.1 | XP_016860202.1 | 34 | 4712 | |||
XM_017004714.1 | XP_016860203.1 | 34 | 4648 | |||
GenBank mRNA | AJ278357.1 | - | 35 | 4620 | ||
AK023807.1 | - | 22 | 2831 | |||
AL832013.1 | - | 24 | 2961 | |||
BC104902.1 | - | 35 | 4407 | 8143905 | ||
BC104904.1 | - | 35 | 4407 | 8143907 |
Gene Symbol: LOC102724642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533233.2 | XP_011531535.2 | 18 | 2369 |