Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022761.2 | NP_073598.1 | 4 | 814 | ||
XM_011542957.2 | XP_011541259.1 | 5 | 979 | |||
XM_011542958.2 | XP_011541260.1 | 4 | 583 | |||
XM_017018129.1 | XP_016873618.1 | 3 | 468 | |||
GenBank mRNA | AJ250229.1 | - | 4 | 512 | ||
AK027152.1 | - | 4 | 497 | |||
BC020628.1 | - | 4 | 533 | 4656696 |