Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022782.3 | NP_073619.3 | 18, 19 | 3084 | ||
NR_103517.1 | 18, 19 | 2823 | ||||
XM_011537740.2 | XP_011536042.1 | 18, 19 | 2988 | |||
XM_011537741.2 | XP_011536043.1 | 18, 19 | 2988 | |||
XM_017018673.1 | XP_016874162.1 | 18, 19 | 3082 | |||
XM_017018674.1 | XP_016874163.1 | 17, 18 | 2886 | |||
XM_017018675.1 | XP_016874164.1 | 17, 18 | 2890 | |||
XM_017018676.1 | XP_016874165.1 | 17, 18 | 2832 | |||
XM_017018677.1 | XP_016874166.1 | 18, 19 | 2988 | |||
XM_017018678.1 | XP_016874167.1 | 17, 18 | 2808 | |||
XM_017018679.1 | XP_016874168.1 | 19, 20 | 3021 | |||
XM_017018680.1 | XP_016874169.1 | 17, 18 | 2930 | |||
XM_017018681.1 | XP_016874170.1 | 17, 18 | 2843 | |||
GenBank mRNA | AK023016.1 | - | 13, 14 | 1807 | ||
AK091181.1 | - | 14, 15 | 2413 | |||
AK096541.1 | - | 18, 19 | 2696 | |||
AL096751.1 | - | 12, 13 | 1744 | |||
BC130440.1 | - | 14, 15 | 2366 | 40146471 | ||
BC144016.1 | - | 13, 14 | 2240 | |||
BC144017.1 | - | 13, 14 | 2018 |