Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002738.6 | NP_002729.2 | 17 | 2151 | ||
GenBank mRNA | BC036472.1 | - | 17 | 2111 | 5260547 | |
M13975.1 | - | 17 | 2018 | |||
X07109.1 | - | 17 | 2090 |