Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 3 | 416 | ||
NM_001258373.1 | NP_001245302.1 | 4 | 519 | |||
NM_022827.3 | NP_073738.2 | 4 | 464 | |||
GenBank mRNA | AK025000.1 | - | 3 | 313 | ||
AK092731.1 | - | 4 | 449 | |||
AK125807.1 | - | 6 | 1194 | |||
AK225189.1 | - | 3 | 313 | |||
AK302850.1 | - | 3 | 363 | |||
AK308616.1 | - | 4 | 589 | |||
AK310416.1 | - | 3 | 308 | |||
BC025255.1 | - | 4 | 352 | 5019010 | ||
BC065526.1 | - | 4 | 406 | 6149633 | ||
BC111029.1 | - | 3 | 321 | 5245538 | ||
BM787432.1 | - | 3 | 379 | |||
CR936769.1 | - | 3 | 334 | |||
DA295930.1 | - | 4 | 479 | |||
DA489234.1 | - | 4 | 464 | |||
DQ238597.1 | - | 3 | 382 | |||
GQ472220.1 | - | 3 | 321 |