Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040097.1 | NP_001035186.1 | 4 | 597 | ||
NM_001040098.1 | NP_001035187.1 | 4 | 513 | |||
NM_001040099.1 | NP_001035188.1 | 3 | 666 | |||
NM_023948.4 | NP_076438.1 | 3 | 696 | |||
XM_005250529.4 | XP_005250586.1 | 4 | 518 | |||
XM_005250531.3 | XP_005250588.1 | 4 | 543 | |||
GenBank mRNA | BC005042.1 | - | 3 | 625 | 3689026 | |
BC011653.2 | - | 3 | 677 | 4299899 | ||
BC028475.2 | - | 3 | 601 | |||
BQ923161.1 | - | 571 |