Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286561.1 | NP_001273490.1 | 16 | 4443 | ||
NM_006185.3 | NP_006176.2 | 15 | 4345 | |||
XM_006718564.1 | XP_006718627.1 | 14 | 4160 | |||
XM_011545054.1 | XP_011543356.1 | 14 | 4160 | |||
XM_011545055.1 | XP_011543357.1 | 15 | 4338 | |||
XM_011545056.2 | XP_011543358.1 | 16 | 4332 | |||
XM_011545057.2 | XP_011543359.1 | 16 | 4322 | |||
XM_011545058.1 | XP_011543360.1 | 14 | 4359 | |||
XM_011545059.2 | XP_011543361.1 | 16 | 4324 | |||
XM_011545060.2 | XP_011543362.1 | 16 | 4318 | |||
XM_011545061.2 | XP_011543363.1 | 14 | 4178 | |||
XM_011545062.2 | XP_011543364.1 | 14 | 4191 | |||
XM_011545063.2 | XP_011543365.1 | 14 | 4158 | |||
XM_011545064.1 | XP_011543366.1 | 14 | 4145 | |||
XM_011545065.1 | XP_011543367.1 | 14 | 4160 | |||
XM_011545066.1 | XP_011543368.1 | 14 | 4160 | |||
XM_017017830.1 | XP_016873319.1 | 15 | 4200 | |||
XM_017017831.1 | XP_016873320.1 | 14 | 4196 | |||
GenBank mRNA | AB208841.1 | - | 1 | 2460 | ||
AB210007.1 | - | 15 | 4146 | |||
BC008345.1 | - | 1 | 541 | |||
BC013023.2 | - | 1 | 1289 | 3506457 | ||
Z11583.1 | - | 15 | 4163 | |||
Z11584.1 | - | 16 | 4265 | |||
Z14227.1 | - | 1 | 2180 | |||
Z14228.1 | - | 1 | 1085 |