Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024052.4 | NP_076957.3 | 2, 3 | 663 | ||
XM_005256799.4 | XP_005256856.1 | 2, 3 | 550 | |||
XM_005256800.1 | XP_005256857.1 | 2, 3 | 550 | |||
XM_011524008.1 | XP_011522310.1 | 2, 3 | 550 | |||
GenBank mRNA | AK074284.1 | - | 2, 3 | 538 | ||
AK127580.1 | - | 2, 3 | 350 | |||
AK315083.1 | - | 2, 3 | 550 | |||
BC000636.2 | - | 2, 3 | 324 | |||
BC041829.1 | - | 2, 3 | 668 | 5268071 |