Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 20 | 2170 | ||
NM_023015.4 | NP_075391.3 | 19 | 2511 | |||
XM_005245459.2 | XP_005245516.1 | 19 | 2528 | |||
XM_005245461.2 | XP_005245518.1 | 13 | 1201 | |||
XM_006711490.3 | XP_006711553.1 | 20 | 1981 | |||
XM_011509906.2 | XP_011508208.1 | 19 | 1893 | |||
XM_017002132.1 | XP_016857621.1 | 9 | 769 | |||
GenBank mRNA | AK025572.1 | - | 5 | 332 | ||
AK074979.1 | - | 12 | 1209 | |||
AK225294.1 | - | 5 | 332 | |||
AK290025.1 | - | 20 | 2164 | |||
AK298746.1 | - | 7 | 661 | |||
AK304874.1 | - | 13 | 1600 | |||
AL080172.1 | - | 14 | 1410 | |||
AL832133.1 | - | 18 | 3546 | |||
BC025254.1 | - | 3 | 171 | |||
BC054513.1 | - | 6 | 547 | |||
BC073985.1 | - | 3 | 183 | |||
BC098431.1 | - | 5 | 371 | |||
BC105092.1 | - | 19 | 1974 | 8144095 | ||
BC105094.1 | - | 19 | 1974 | 8144097 | ||
BC116458.1 | - | 19 | 2506 | 5752962 | ||
BX640786.1 | - | 19 | 2497 | |||
BX640950.1 | - | 19 | 2512 | |||
CR749212.1 | - | 17 | 4038 | |||
CR749376.1 | - | 18 | 4163 |