Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171251.2 | NP_001164722.1 | 3 | 554 | ||
NM_001321310.1 | NP_001308239.1 | 3 | 554 | |||
NM_001321311.1 | NP_001308240.1 | 3 | 554 | |||
NM_024032.4 | NP_076937.2 | 3 | 554 | |||
XM_011525187.1 | XP_011523489.1 | 3 | 404 | |||
XM_011525188.1 | XP_011523490.1 | 3 | 404 | |||
XM_011525189.1 | XP_011523491.1 | 3 | 404 | |||
XM_011525190.1 | XP_011523492.1 | 3 | 404 | |||
XM_011525191.2 | XP_011523493.1 | 3 | 404 | |||
XM_011525192.1 | XP_011523494.1 | 3 | 404 | |||
XM_011525193.1 | XP_011523495.1 | 3 | 404 | |||
XM_011525194.1 | XP_011523496.1 | 3 | 404 | |||
XM_011525195.1 | XP_011523497.1 | 3 | 404 | |||
XM_017025031.1 | XP_016880520.1 | 3 | 577 | |||
XM_017025032.1 | XP_016880521.1 | 3 | 576 | |||
GenBank mRNA | AK291924.1 | - | 3 | 548 | ||
AL834295.1 | - | 3 | 498 | |||
BC000121.1 | - | 3 | 434 | 3352851 | ||
BI829278.1 | - | 3 | 550 |