Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002876.2 | NP_001002876.1 | 4 | 384 | ||
NM_001304370.1 | NP_001291299.1 | 3 | 497 | |||
NM_001304371.1 | NP_001291300.1 | 4 | 384 | |||
NM_001304372.1 | NP_001291301.1 | 3 | 582 | |||
NM_001304373.1 | NP_001291302.1 | 3 | 497 | |||
NM_024053.4 | NP_076958.1 | 4 | 384 | |||
XM_011530368.2 | XP_011528670.1 | 4 | 371 | |||
GenBank mRNA | AK123479.1 | - | 4 | 373 | ||
AL160131.1 | - | 4 | 332 | |||
BC000705.2 | - | 4 | 331 | 3349507 | ||
BC007495.2 | - | 4 | 344 | 2966541 | ||
BE789455.1 | - | 3 | 429 | |||
BE797487.1 | - | 4 | 338 | |||
BM045658.1 | - | 3 | 557 | |||
BQ652721.1 | - | 3 | 334 | |||
BQ922403.1 | - | 4 | 317 | |||
BU556693.1 | - | 4 | 384 | |||
BX332140.2 | - | 3 | 435 | |||
BX354019.2 | - | 3 | 431 | |||
BX417558.2 | - | 3 | 436 | |||
CR456399.1 | - | 4 | 357 | |||
CR457392.1 | - | 4 | 296 |