Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324475.1 | NP_001311404.1 | 31 | 3425 | ||
NM_023015.4 | NP_075391.3 | 30 | 3766 | |||
XM_005245459.2 | XP_005245516.1 | 29 | 3981 | |||
XM_005245461.2 | XP_005245518.1 | 24 | 2456 | |||
XM_006711490.3 | XP_006711553.1 | 30 | 3434 | |||
XM_011509906.2 | XP_011508208.1 | 29 | 3346 | |||
XM_017002132.1 | XP_016857621.1 | 19 | 2222 | |||
GenBank mRNA | AK025572.1 | - | 16 | 1587 | ||
AK055174.1 | - | 9 | 2149 | |||
AK074979.1 | - | 23 | 2464 | |||
AK094402.1 | - | 1 | 698 | |||
AK126799.1 | - | 10 | 2304 | |||
AK225294.1 | - | 16 | 1587 | |||
AK290025.1 | - | 31 | 3419 | |||
AK304874.1 | - | 23 | 3053 | |||
AL080172.1 | - | 25 | 2665 | |||
AL832133.1 | - | 29 | 4801 | |||
BC025254.1 | - | 14 | 1426 | |||
BC054513.1 | - | 17 | 1802 | |||
BC073985.1 | - | 14 | 1438 | |||
BC098431.1 | - | 16 | 1626 | |||
BC116458.1 | - | 30 | 3761 | 5752962 | ||
BX640786.1 | - | 30 | 3752 | |||
BX640950.1 | - | 30 | 3767 | |||
CR749212.1 | - | 28 | 5293 | |||
CR749376.1 | - | 29 | 5418 |