Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVRIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024070.3 | NP_076975.2 | 6 | 1320 | ||
XM_011516575.2 | XP_011514877.1 | 7 | 1336 | |||
GenBank mRNA | BC001129.2 | - | 5 | 1012 | ||
BC032113.2 | - | 3 | 711 | |||
BC073861.1 | - | 5 | 983 | |||
BC107873.1 | - | 7 | 1345 | 6452536 | ||
BC112090.1 | - | 5 | 1048 | |||
BC112092.1 | - | 5 | 1049 |