Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024097.3 | NP_077002.2 | 2, 3 | 261 | ||
GenBank mRNA | BC001508.2 | - | 2, 3 | 237 | 2959109 | |
BC001711.1 | - | 2, 3 | 237 | 3543543 | ||
DA684965.1 | - | 2, 3 | 261 |