Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024108.2 | NP_077013.1 | 1, 2 | 159 | ||
XM_017027295.1 | XP_016882784.1 | 1, 2 | 218 | |||
GenBank mRNA | BC001907.1 | - | 1, 2 | 143 | 3534111 | |
BC004450.1 | - | 1, 2 | 144 | 2820830 |