Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024323.3 | NP_077299.3 | 6 | 1394 | ||
XM_006722885.3 | XP_006722948.2 | 6 | 1767 | |||
XM_011528286.2 | XP_011526588.1 | 5 | 1556 | |||
XM_011528287.2 | XP_011526589.1 | 5 | 1216 | |||
XM_017027300.1 | XP_016882789.1 | 5 | 1554 | |||
XM_017027301.1 | XP_016882790.1 | 5 | 1418 | |||
GenBank mRNA | AK097431.1 | - | 6 | 1394 | ||
BC002891.2 | - | 1 | 520 | |||
BC012945.1 | - | 2 | 970 | |||
BC119719.1 | - | 5 | 1362 | 40117748 |