Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323640.1 | NP_001310569.1 | 3 | 1509 | ||
NM_024063.2 | NP_076968.2 | 3 | 1509 | |||
NR_027635.1 | 3 | 1509 | ||||
NR_136645.1 | 3 | 1516 | ||||
NR_136646.1 | 4 | 1704 | ||||
NR_136647.1 | 4 | 1671 | ||||
NR_136648.1 | 4 | 1704 | ||||
GenBank mRNA | AK023232.1 | - | 3 | 1455 | ||
AK225463.1 | - | 3 | 1455 | |||
AK291457.1 | - | 3 | 1462 | |||
BC000981.2 | - | 3 | 1442 | 2985725 | ||
BC051861.2 | - | 3 | 1454 |