Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310.3 | NP_001301.1 | 2, 3 | 549 | ||
GenBank mRNA | AB451461.1 | - | 1, 2 | 209 | ||
AF039081.1 | - | 2, 3 | 485 | |||
BC104873.1 | - | 2, 3 | 393 | 8143876 | ||
BC106052.1 | - | 2, 3 | 490 | 6150812 | ||
BC112157.1 | - | 2, 3 | 393 | 8327625 | ||
BY795169.2 | - | 2, 3 | 549 | |||
CR542250.1 | - | 1, 2 | 209 |