Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024063.2 | NP_076968.2 | 5, 6 | 1953 | ||
NR_136645.1 | 5, 6 | 1960 | ||||
NR_136648.1 | 6, 7 | 2148 | ||||
GenBank mRNA | AK225463.1 | - | 5, 6 | 1899 | ||
BC000981.2 | - | 5, 6 | 1886 | 2985725 | ||
BC051861.2 | - | 5, 6 | 1898 |