Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122835.2 | NP_001116307.2 | 12, 13 | 1612 | ||
NM_024525.4 | NP_078801.3 | 14, 15 | 1771 | |||
XM_005273260.1 | XP_005273317.1 | 14, 15 | 1771 | |||
XM_005273261.2 | XP_005273318.1 | 13, 14 | 1678 | |||
XM_005273262.1 | XP_005273319.1 | 12, 13 | 1612 | |||
XM_005273264.2 | XP_005273321.1 | 12, 13 | 1543 | |||
XM_006711814.2 | XP_006711877.1 | 10, 11 | 1384 | |||
XM_006711815.2 | XP_006711878.1 | 7, 8 | 2778 | |||
XM_011544275.2 | XP_011542577.1 | 11, 12 | 1450 | |||
XM_011544276.2 | XP_011542578.1 | 7, 8 | 1014 | |||
XM_017002321.1 | XP_016857810.1 | 13, 14 | 1678 | |||
XM_017002322.1 | XP_016857811.1 | 12, 13 | 1543 | |||
XM_017002323.1 | XP_016857812.1 | 10, 11 | 1346 | |||
XM_017002324.1 | XP_016857813.1 | 10, 11 | 1384 | |||
XM_017002325.1 | XP_016857814.1 | 14, 15 | 4976 | |||
XM_017002326.1 | XP_016857815.1 | 9, 10 | 4356 | |||
GenBank mRNA | AK026237.1 | - | 1, 2 | 78 | ||
AK074365.1 | - | 5, 6 | 679 | |||
AK075386.1 | - | 14, 15 | 1737 | |||
AK075561.1 | - | 12, 13 | 1591 | |||
AK309725.1 | - | 10, 11 | 1430 | |||
AL832286.1 | - | 14, 15 | 1506 | |||
AL834453.1 | - | 5, 6 | 740 | |||
BC042683.1 | - | 5, 6 | 658 |