Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040610.2 | NP_001035700.1 | 6, 7 | 879 | ||
NM_001322844.1 | NP_001309773.1 | 7, 8 | 947 | |||
NM_001322845.1 | NP_001309774.1 | 8, 9 | 1274 | |||
NM_024580.5 | NP_078856.4 | 8, 9 | 1032 | |||
NR_136410.1 | 8, 9 | 1032 | ||||
XM_011522000.1 | XP_011520302.1 | 6, 7 | 703 | |||
GenBank mRNA | AK023181.1 | - | 2, 3 | 184 | ||
AK056656.1 | - | 8, 9 | 962 | |||
AK300348.1 | - | 8, 9 | 942 | |||
BX538332.1 | - | 6, 7 | 810 |