Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300747.1 | NP_001287676.1 | 2 | 366 | ||
NM_001300748.1 | NP_001287677.1 | 2 | 366 | |||
NM_024587.3 | NP_078863.2 | 2 | 366 | |||
XM_011542138.2 | XP_011540440.1 | 2 | 585 | |||
GenBank mRNA | AA490195.1 | - | 2 | 161 | ||
AK026006.1 | - | 2 | 173 | |||
AK225405.1 | - | 2 | 173 | |||
AK295356.1 | - | 1 | 59 | |||
AK310016.1 | - | 2 | 400 | |||
BC064520.1 | - | 2 | 185 | 5582377 | ||
BM806701.1 | - | 3 | 247 | |||
CR457348.1 | - | 2 | 149 | |||
HY029536.1 | - | 2 | 200 | |||
HY108001.1 | - | 2 | 368 |