Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253826.1 | NP_001240755.1 | 2, 3 | 877 | ||
NM_001253827.1 | NP_001240756.1 | 2, 3 | 472 | |||
XM_011533105.1 | XP_011531407.1 | 2, 3 | 226 | |||
GenBank mRNA | AK022753.1 | - | 2, 3 | 298 | ||
AK122747.1 | - | 2, 3 | 456 |