Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024590.3 | NP_078866.3 | 2 | 2428 | ||
XM_005263212.4 | XP_005263269.1 | 2 | 2386 | |||
XM_011532238.2 | XP_011530540.1 | 3 | 1765 | |||
XM_011532239.2 | XP_011530541.1 | 4 | 1303 | |||
XM_017008592.1 | XP_016864081.1 | 3 | 2468 | |||
XM_017008593.1 | XP_016864082.1 | 6 | 3023 | |||
XM_017008594.1 | XP_016864083.1 | 5 | 2590 | |||
XM_017008595.1 | XP_016864084.1 | 6 | 2703 | |||
XM_017008596.1 | XP_016864085.1 | 6 | 2683 | |||
XM_017008597.1 | XP_016864086.1 | 6 | 2714 | |||
XM_017008598.1 | XP_016864087.1 | 4 | 2540 | |||
XM_017008599.1 | XP_016864088.1 | 4 | 2549 | |||
XM_017008600.1 | XP_016864089.1 | 3 | 1253 | |||
GenBank mRNA | AK027201.1 | - | 1 | 453 | ||
AK307532.1 | - | 4 | 2070 | |||
AM049401.1 | - | 2 | 1702 | |||
AY358647.1 | - | 2 | 2429 | |||
AY875938.1 | - | 2 | 1540 | |||
BC032010.1 | - | 1 | 505 | |||
BC033156.1 | - | 2 | 1332 | |||
BC089445.1 | - | 2 | 1798 | |||
BC132879.1 | - | 2 | 1628 | 40146901 | ||
BC132881.1 | - | 2 | 1628 | 40146903 | ||
BC144265.1 | - | 2 | 1628 | 9052785 | ||
BC151213.1 | - | 3 | 2644 |