Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135109.2 | NP_001128581.1 | 2 | 1850 | ||
NM_001206671.2 | NP_001193600.1 | 6 | 2396 | |||
NM_001206672.2 | NP_001193601.1 | 4 | 2153 | |||
NM_024557.4 | NP_078833.3 | 6 | 2393 | |||
NR_045405.1 | 7 | 2445 | ||||
XM_006718317.3 | XP_006718380.1 | 7 | 2274 | |||
XM_006718318.3 | XP_006718381.1 | 7 | 2187 | |||
XM_017018287.1 | XP_016873776.1 | 6 | 2525 | |||
XM_017018288.1 | XP_016873777.1 | 7 | 2522 | |||
XM_017018289.1 | XP_016873778.1 | 7 | 2469 | |||
XM_017018290.1 | XP_016873779.1 | 5 | 2292 | |||
GenBank mRNA | AL832601.2 | - | 2 | 1811 | ||
AY326435.1 | - | 6 | 2342 | |||
BC022455.1 | - | 6 | 2375 | 4792930 | ||
BX647514.1 | - | 3 | 1993 | |||
DQ347704.1 | - | 1 | 1139 | |||
DQ347705.1 | - | 1 | 1138 |