Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3TC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024577.3 | NP_078853.2 | 17 | 4702 | ||
GenBank mRNA | AB075865.1 | - | 18 | 4644 | ||
AK023667.1 | - | 2 | 1773 | |||
AK124854.1 | - | 2 | 2606 | |||
AK127050.1 | - | 7 | 2267 | |||
AK127241.1 | - | 13 | 4071 | |||
AK128209.1 | - | 7 | 3330 |