Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304271.1 | NP_001291200.1 | 3 | 1016 | ||
NM_024585.3 | NP_078861.1 | 3 | 1080 | |||
GenBank mRNA | AK025813.1 | - | 3 | 888 | ||
AK309353.1 | - | 2 | 855 | |||
BC011728.1 | - | 3 | 1045 | 4423125 |