Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018667.3 | NP_061137.1 | 9 | 4749 | ||
XM_005256031.3 | XP_005256088.1 | 9 | 4842 | |||
XM_005256032.3 | XP_005256089.1 | 9 | 4861 | |||
XM_011523207.1 | XP_011521509.1 | 10 | 4842 | |||
XM_011523208.2 | XP_011521510.1 | 8 | 4943 | |||
XM_011523209.2 | XP_011521511.1 | 8 | 4726 | |||
XM_017023405.1 | XP_016878894.1 | 9 | 4668 | |||
XM_017023406.1 | XP_016878895.1 | 10 | 5149 | |||
XM_017023407.1 | XP_016878896.1 | 10 | 4871 | |||
XM_017023408.1 | XP_016878897.1 | 10 | 4825 | |||
GenBank mRNA | AK026246.1 | - | 1 | 907 | ||
BC012727.2 | - | 1 | 1295 |