Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304833.1 | NP_001291762.1 | 7 | 1268 | ||
NM_001304834.1 | NP_001291763.1 | 8 | 1699 | |||
NM_001304835.1 | NP_001291764.1 | 7 | 1819 | |||
NM_001304836.1 | NP_001291765.1 | 7 | 1588 | |||
NM_001304837.1 | NP_001291766.1 | 6 | 1449 | |||
NM_001304838.1 | NP_001291767.1 | 7 | 1506 | |||
NM_024623.2 | NP_078899.1 | 8 | 1655 | |||
GenBank mRNA | AK023553.1 | - | 8 | 1551 | ||
AK094820.1 | - | 7 | 1251 | |||
AK095481.1 | - | 6 | 1357 | |||
BC098119.1 | - | 8 | 1431 | 40024912 | ||
BC098293.1 | - | 8 | 1431 | 40024914 | ||
BC101940.1 | - | 7 | 1238 | 40024915 | ||
BC105637.1 | - | 7 | 1238 | 40024913 | ||
BX640734.1 | - | 8 | 1299 |