Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195483.1 | NP_001182412.1 | 9 | 1467 | ||
NM_024628.5 | NP_078904.3 | 10 | 1528 | |||
GenBank mRNA | AF345197.1 | - | 9 | 1417 | ||
AF390442.1 | - | 9 | 1468 | |||
AK026841.1 | - | 6 | 864 | |||
AK123001.1 | - | 10 | 1482 | |||
AK308484.1 | - | 5 | 785 | |||
BC020506.1 | - | 2 | 430 | |||
BC063528.1 | - | 5 | 751 | |||
BC126158.1 | - | 9 | 1438 | 8991874 | ||
BC126160.1 | - | 9 | 1438 | 8991876 | ||
BX538023.1 | - | 4 | 851 | |||
CR457363.1 | - | 1 | 199 |