Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080412.2 | NP_001073881.2 | 8 | 3794 | ||
XM_005247256.4 | XP_005247313.1 | 2 | 4314 | |||
XM_005247257.1 | XP_005247314.1 | 3 | 2983 | |||
XM_005247258.1 | XP_005247315.1 | 4 | 3423 | |||
XM_005247261.3 | XP_005247318.1 | 3 | 3052 | |||
XM_011512607.1 | XP_011510909.1 | 4 | 3070 | |||
XM_011512608.1 | XP_011510910.1 | 4 | 3134 | |||
XM_011512611.1 | XP_011510913.1 | 4 | 3075 | |||
XM_017006050.1 | XP_016861539.1 | 5 | 3379 | |||
XM_017006051.1 | XP_016861540.1 | 5 | 3574 | |||
XM_017006052.1 | XP_016861541.1 | 4 | 3497 | |||
XM_017006053.1 | XP_016861542.1 | 5 | 3193 | |||
XM_017006054.1 | XP_016861543.1 | 5 | 3294 | |||
XM_017006055.1 | XP_016861544.1 | 5 | 3198 | |||
XM_017006056.1 | XP_016861545.1 | 5 | 3322 | |||
XM_017006057.1 | XP_016861546.1 | 3 | 3574 | |||
XM_017006058.1 | XP_016861547.1 | 5 | 3217 | |||
XM_017006059.1 | XP_016861548.1 | 4 | 3195 | |||
XM_017006060.1 | XP_016861549.1 | 5 | 3232 | |||
XM_017006061.1 | XP_016861550.1 | 5 | 3173 | |||
GenBank mRNA | AK025985.1 | - | 1 | 480 | ||
AK094201.1 | - | 1 | 1100 | |||
AK302568.1 | - | 2 | 2862 | |||
AL833148.1 | - | 1 | 1096 | |||
BC107745.1 | - | 1 | 286 |