Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024649.4 | NP_078925.3 | 17 | 2899 | ||
GenBank mRNA | AF503941.1 | - | 17 | 2899 | ||
AK027243.1 | - | 5 | 1597 | |||
AK095638.1 | - | 15 | 3114 | |||
BC047642.1 | - | 16 | 2976 | 4824649 | ||
BX647612.1 | - | 17 | 2928 |