Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287527.1 | NP_001274456.1 | 9 | 1319 | ||
NM_001287528.1 | NP_001274457.1 | 9 | 1355 | |||
NM_024692.5 | NP_078968.3 | 9 | 1323 | |||
NR_109844.1 | 8 | 1261 | ||||
XM_005264562.2 | XP_005264619.1 | 9 | 1350 | |||
XM_005264563.1 | XP_005264620.1 | 9 | 1355 | |||
XM_006712104.2 | XP_006712167.1 | 9 | 1395 | |||
XM_006712105.3 | XP_006712168.1 | 9 | 1237 | |||
XM_011533109.1 | XP_011531411.1 | 9 | 1103 | |||
XM_011533110.1 | XP_011531412.1 | 8 | 1156 | |||
XM_017004958.1 | XP_016860447.1 | 9 | 1182 | |||
XM_017004959.1 | XP_016860448.1 | 9 | 1237 | |||
GenBank mRNA | AB209042.1 | - | 9 | 2934 | ||
AF433661.1 | - | 9 | 1331 | |||
AK024722.1 | - | 6 | 766 | |||
AK057267.1 | - | 8 | 1262 | |||
AK225074.1 | - | 6 | 766 | |||
AK291595.1 | - | 9 | 1400 | |||
AK302325.1 | - | 9 | 1305 | |||
AK304384.1 | - | 8 | 1149 | |||
AL834443.1 | - | 9 | 1297 | |||
BC013862.1 | - | 9 | 1303 | |||
BC015310.1 | - | 7 | 929 | |||
BC126167.1 | - | 8 | 1093 | 8991883 | ||
BC136331.1 | - | 8 | 1093 | 9020318 |