Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024715.1 | - | 10 | 1180 | ||
AK302636.1 | - | 22 | 2725 | |||
AK303034.1 | - | 22 | 2799 | |||
AL833446.1 | - | 14 | 6540 | |||
BC132771.1 | - | 16 | 2042 | |||
BC141834.1 | - | 22 | 2766 | 40148850 | ||
BC142655.1 | - | 22 | 2766 | 40148060 |