Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284403.1 | NP_001271332.1 | 7 | 653 | ||
NM_001284404.1 | NP_001271333.1 | 8 | 783 | |||
NM_001284405.1 | NP_001271334.1 | 9 | 901 | |||
NM_024754.4 | NP_079030.3 | 10 | 980 | |||
XM_005248601.2 | XP_005248658.1 | 9 | 860 | |||
XM_005248603.2 | XP_005248660.1 | 9 | 836 | |||
XM_017009866.1 | XP_016865355.1 | 9 | 841 | |||
XM_017009867.1 | XP_016865356.1 | 8 | 753 | |||
XM_017009868.1 | XP_016865357.1 | 8 | 722 | |||
GenBank mRNA | AK022660.1 | - | 8 | 742 | ||
AK056761.1 | - | 10 | 1644 | |||
AK298750.1 | - | 7 | 644 | |||
AK303639.1 | - | 8 | 752 | |||
BC018720.1 | - | 10 | 956 | 4552964 | ||
CR457290.1 | - | 6 | 508 |