Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024761.4 | NP_079037.3 | 2 | 807 | ||
GenBank mRNA | AJ580636.1 | - | 1 | 311 | ||
AK023266.1 | - | 2 | 736 | |||
AL832572.2 | - | 2 | 701 | |||
BC033027.1 | - | 2 | 719 | 5268356 | ||
CR457307.1 | - | 1 | 311 |