Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318006.1 | NP_001304935.1 | 4 | 548 | ||
NM_001318007.1 | NP_001304936.1 | 4 | 442 | |||
NM_001318008.1 | NP_001304937.1 | 3 | 346 | |||
NM_024792.2 | NP_079068.1 | 5 | 644 | |||
GenBank mRNA | AF477201.1 | - | 5 | 632 | ||
AK025935.1 | - | 5 | 583 | |||
AK125146.1 | - | 4 | 849 | |||
AK292065.1 | - | 5 | 583 | |||
BC009729.1 | - | 2 | 137 | |||
BC026023.1 | - | 4 | 510 | 4548729 | ||
BX389422.1 | - | 3 | 189 | |||
CN259418.1 | - | 3 | 355 |