Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286429.1 | NP_001273358.1 | 5 | 810 | ||
NM_024817.2 | NP_079093.2 | 10 | 1761 | |||
XM_006720692.3 | XP_006720755.1 | 11 | 1844 | |||
XM_011522043.2 | XP_011520345.1 | 5 | 2529 | |||
XM_011522044.2 | XP_011520346.1 | 6 | 2170 | |||
XM_017022582.1 | XP_016878071.1 | 6 | 905 | |||
XM_017022583.1 | XP_016878072.1 | 7 | 845 | |||
XM_017022584.1 | XP_016878073.1 | 6 | 812 | |||
XM_017022585.1 | XP_016878074.1 | 6 | 727 | |||
XM_017022586.1 | XP_016878075.1 | 6 | 2040 | |||
GenBank mRNA | AK023772.1 | - | 3 | 337 | ||
AK131551.1 | - | 11 | 1816 | |||
AK299056.1 | - | 5 | 810 | |||
AK303870.1 | - | 5 | 784 | |||
BC140868.1 | - | 10 | 1715 | 9021736 | ||
BX648716.1 | - | 5 | 775 | |||
BX648747.1 | - | 5 | 618 |