Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024743.3 | NP_079019.3 | 4 | 1054 | ||
NR_024010.1 | 5 | 1151 | ||||
XM_011532247.2 | XP_011530549.1 | 4 | 1081 | |||
GenBank mRNA | AK025587.1 | - | 4 | 859 | ||
AK225308.1 | - | 4 | 859 | |||
AK293381.1 | - | 5 | 722 | |||
AY358727.1 | - | 4 | 1049 | |||
AY542891.1 | - | 4 | 1054 | |||
BC130533.1 | - | 4 | 1047 | 40146564 | ||
BP262099.1 | - | 5 | 449 |