Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024769.3 | NP_079045.1 | 7 | 1798 | ||
GenBank mRNA | AK026068.1 | - | 7 | 1774 | ||
AY326422.1 | - | 7 | 1622 | |||
AY358340.1 | - | 7 | 1622 | |||
BC009371.2 | - | 7 | 1444 | 4122648 |