Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024786.2 | NP_079062.1 | 6 | 1252 | ||
XM_017009869.1 | XP_016865358.1 | 7 | 1409 | |||
XM_017009870.1 | XP_016865359.1 | 9 | 3220 | |||
XM_017009871.1 | XP_016865360.1 | 9 | 3169 | |||
XM_017009872.1 | XP_016865361.1 | 7 | 1244 | |||
XM_017009873.1 | XP_016865362.1 | 3 | 1226 | |||
XM_017009874.1 | XP_016865363.1 | 3 | 2106 | |||
XM_017009875.1 | XP_016865364.1 | 3 | 2053 | |||
XM_017009876.1 | XP_016865365.1 | 3 | 1020 | |||
XM_017009877.1 | XP_016865366.1 | 3 | 594 | |||
XM_017009878.1 | XP_016865367.1 | 3 | 551 | |||
GenBank mRNA | AK023215.1 | - | 6 | 1252 | ||
AK024233.1 | - | 3 | 1931 | |||
BC032000.1 | - | 6 | 871 | 4750082 |
Gene Symbol: ZDHHC11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010110.1 | XP_016865599.1 | 10 | 1517 | ||
XM_017010111.1 | XP_016865600.1 | 9 | 1344 | |||
XM_017010112.1 | XP_016865601.1 | 9 | 1921 | |||
XM_017010113.1 | XP_016865602.1 | 10 | 2043 | |||
XM_017010114.1 | XP_016865603.1 | 10 | 4804 | |||
XM_017010115.1 | XP_016865604.1 | 9 | 1869 | |||
XM_017010116.1 | XP_016865605.1 | 8 | 2269 | |||
XM_017010117.1 | XP_016865606.1 | 8 | 2104 | |||
XM_017010118.1 | XP_016865607.1 | 8 | 2261 | |||
XM_017010119.1 | XP_016865608.1 | 8 | 2216 | |||
XM_017010120.1 | XP_016865609.1 | 9 | 3217 | |||
XM_017010122.1 | XP_016865611.1 | 9 | 3214 | |||
XM_017010123.1 | XP_016865612.1 | 8 | 2018 | |||
XM_017010124.1 | XP_016865613.1 | 9 | 2028 | |||
XM_017010125.1 | XP_016865614.1 | 5 | 1593 |